Learn More
PADI4 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL488-17373-100UL
Beschreibung
The PADI4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PADI4. This antibody recognizes human, mouse, rat antigen. The PADI4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spezifikation
| PADI4 | |
| 1000 μg/mL | |
| HL 60 PAD, PAD, PAD4, PADI4, PADI5, PDI4, PDI5, Peptidylarginine deiminase IV | |
| 100 μL | |
| 23569 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |