missing translation for 'onlineSavingsMsg'
Learn More

P4HA2 Mouse anti-Human, Mouse, Rat, Clone: 3A9A4, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66604-1-IG-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838635

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 24-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.
Spezifikation

Spezifikation

P4HA2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15460, Q60716
P4ha2
P4HA2 Fusion Protein Ag4714
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
3A9A4
Unconjugated
P4HA2
4 PH alpha 2, P4HA2
Mouse
Protein G
RUO
18452, 360526, 8974
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote