Learn More
p21/CIP1/CDKN1A Antibody (AC8.3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NB100-2750IR
Artikelnummer. 18063656
  • 622.00 CHF / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
p21/CIP1/CDKN1A Monoclonal specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
p21/CIP1/CDKN1A
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CDKN1A
His-tagged human p21/CIP1/CDKN1A expressed in bacleva virus.
0.1 mL
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair
1026
Store at 4C in the dark.
IgG1
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
AC8.3
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Mouse
Protein A purified
RUO
Primary
Human
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen