missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals p21-aktivierter Kinase-Inhibitor III, IPA-3
Click to view available options
Menge:
5 mg
10 mg
Beschreibung
Acts as an allosteric inhibitor of Pak1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Menge | 10 mg |
Beschreibung | Gelb |
Inhalt und Lagerung | -30°bis -10 °C |
Zur Verwendung mit (Anwendung) | Wirkt als allosterischer Inhibitor von Pak1 |
Formelmasse | 350,50g/mol |
Empfindlichkeit | Lichtempfindlich |
Löslichkeit | Löslich in DMSO oder Ethanol; unlöslich in Wasser |
Produkttyp | Signaltransduktionsreagenz |
Form | Fest |
InChI-Schlüssel | RFAXLXKIAKIUDT-UHFFFAOYSA-N |
Mehr anzeigen |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'