missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracyclin-Dihydrat, TRC

Product Code. 30411252
Change view
Click to view available options
Menge:
50 mg
1 g
Unit Size:
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30411252 1 g -
30419140 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30411252 Supplier Toronto Research Chemicals Supplier No. TRCO8775011G
 Eingeschränkter Artikel
Verfügbar ab: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-64-6
Summenformel C22 H24 N2 O9 . 2 H2 O
Molekulargewicht (g/mol) 496.46
Synonym Oxytetracycline dihydrate, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, hydrate (1:2), (4S,4aR,5S,5aR,6S,12aS)-, 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, dihydrate (8CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, dihydrate, (4S,4aR,5S,5aR,6S,12aS)- (9CI), 2-Naphthacenecarboxamide, 4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-, dihydrate, [4S-(4α,4aα,5α,5aα,6β,12aα)]-, (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,6,10,12,12a-hexahydroxy-6-methyl-1,11-dioxo-2-naphthacenecarboxamide dihydrate, 5-Hydroxytetracycline dihydrate, Nitox
IUPAC-Name (4S,4aR,5S,5aR,6S,12aS)-4-(Dimethylamino)-3,5,6,10,12,12a-Hexahydroxy-6-methyl-1,11-dioxo-4,4a,5,5a-Tetrahydrotracene-2-Carboxamid; Dihydrat
SMILES O.O.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N)C(=O)c4c(O)cccc4[C@@]3(C)O

Specifications

Chemischer Name oder Material Oxytetracycline dihydrate
Menge 1 g
InChI-Formel InChI=1S/C22H24N2O9.2H2O/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);2*1H2/t12-,13-,14+,17+,21-,22+;;/m1../s1
Formelmasse 496.1693
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.