Learn More
Chemsplash OverBoot with White PVC Sole, Elasticated Ankle and Top
GREENER_CHOICE
Artikelnummer. p-200105901
Click to view available options
Größe:
X-Large
2X-Large
Packungsgröße:
200 Paare
Artikelnummer. 30058443
Marke: Chemsplash 2565.400.007
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
The Non-Slip Overboot is elasticated at the calf and ankle and has a non-slip white PVC sole.
Chemsplash Non-Slip Overboot
  • Chemsplash 55GSM Type PB 6B Material
  • Elasticated Top and Ankles
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • White PVC Sole
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Non-Slip Overboot is made from Chemsplash fabric which is soft and microporous. It is strong and resistant to chemical splash and dust. This overboot is elasticated at the calf and ankle and has a non-slip white PVC sole. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Spezifikation
Größe X-Large
Material Microporous Fabric
Latex Free Yes
Farbe White
Einwegprodukt Single-use
Sterilität Non-sterile
Verschlusstyp Elastic Top
Sohlenart Anti-slip
Widerstand Liquid
Produkttyp Controlled Environments Boot Covers
Cleanroom Class Class 100-1000 (ISO 5-6)