missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin receptor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | Orexin receptor 1 |
---|---|
Konzentration | 0.22 mg/mL |
Anwendungen | Flow Cytometry, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16838444
|
Proteintech
18370-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16828444
|
Proteintech
18370-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.Spezifikation
Orexin receptor 1 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43613, P56718, P58307 | |
230777, 25593, 3061 | |
Orexin receptor 1 Fusion Protein Ag12269 | |
Primary | |
-20°C | |
HCRTR1 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HCRTR1, hypocretin (orexin) receptor 1, Hypocretin receptor type 1, Orexin receptor 1, Orexin receptor type 1, Ox 1 R, Ox1 R, OX1R | |
Hcrtr1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |