Learn More
Thermo Scientific Chemicals Orexin B, Mensch
für zell- und molekularbiologische Anwendungen.
Marke: Thermo Scientific Chemicals J65110.M¬
Additional Details : Gewicht : 0.00001kg
Beschreibung
Orexin is a neuropeptide that modulates the energy and metabolism balance. The effects of orexin are mediated by one of the two G protein-coupled receptors, orexin receptor 1 or 2. OXRs are expressed in oxinergic projection areas of the brain and regulate food intake by reducing systemic and brain signals that drive over-eating. Synthetic peptide corresponding to the C-terminus of human orexin receptor 1, conjugated to KLH. The immunizing peptide has 70% homology with the rat and mouse gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Human | |
-30°C to -10°C | |
2899.33 | |
1 mg | |
Light sensitive |
HPLC | |
Lyophilisiertes Pulver | |
Orexin B, human | |
RUO - research use only | |
Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2 |
Nur für Forschungszwecke