Learn More
Thermo Scientific Chemicals Orexin B, Mensch
für zell- und molekularbiologische Anwendungen.
Marke:  Thermo Scientific Chemicals J65110.M¬
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15446729
  • 285.76 CHF / 1 Milligramm
Verfügbar ab: 18-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Orexin is a neuropeptide that modulates the energy and metabolism balance. The effects of orexin are mediated by one of the two G protein-coupled receptors, orexin receptor 1 or 2. OXRs are expressed in oxinergic projection areas of the brain and regulate food intake by reducing systemic and brain signals that drive over-eating. Synthetic peptide corresponding to the C-terminus of human orexin receptor 1, conjugated to KLH. The immunizing peptide has 70% homology with the rat and mouse gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
Human
-30°C to -10°C
2899.33
1 mg
Light sensitive
HPLC
Lyophilisiertes Pulver
Orexin B, human
RUO - research use only
Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS) Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
Nur für Forschungszwecke