Learn More
OMA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563067
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YLDRLIPQAL KIREMCNCPP LSNPDPRLLF KLSTKHFLEE SEKEDLNITK KQKMDTLPIQ KQEQIPLTYI VEKRTGS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
OMA1 (Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family. It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants, and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.Spezifikation
OMA1 | |
Polyclonal | |
Unconjugated | |
OMA1 | |
2010001O09Rik; DAB1; metalloendopeptidase OMA1, mitochondrial; metalloprotease related protein 1; metalloprotease-related protein 1; MPRP1; MPRP-1; Oma1; OMA1 homolog, zinc metallopeptidase; OMA1 homolog, zinc metallopeptidase (S. cerevisiae); OMA1 zinc metallopeptidase; OMA1 zinc metallopeptidase homolog; overlapping activity with M-AAA protease; overlapping with the m-AAA protease 1 homolog; peptidase; RGD1304821; YKR087C; zinc metallopeptidase homolog; zinc metallopeptidase OMA1; ZMPOMA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115209 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96E52 | |
OMA1 | |
Recombinant protein corresponding to Human OMA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only