missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Olmesartan-Medoxomil

Catalog Number p-7214419
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
144689-63-4
UQGKUQLKSCSZGY-UHFFFAOYSA-N
(5-Methyl-2-Oxo-2H-1,3-Dioxol-4-yl)methyl 4-(2-Hydroxypropan-2-yl)-2-Propyl-1-{[2'-(2H-1,2,3,4-Tetrazol-5-yl)-[1,1'-Biphenyl]-4-yl]Methyl}-1H-Imidazol-5-Carboxylat
CCCC1=NC(=C(N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)C(=O)OCC1=C(C)OC(=O)O1)C(C)(C)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
144689-63-4
UQGKUQLKSCSZGY-UHFFFAOYSA-N
(5-Methyl-2-Oxo-2H-1,3-Dioxol-4-yl)methyl 4-(2-Hydroxypropan-2-yl)-2-Propyl-1-{[2'-(2H-1,2,3,4-Tetrazol-5-yl)-[1,1'-Biphenyl]-4-yl]Methyl}-1H-Imidazol-5-Carboxylat
CCCC1=NC(=C(N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NNN=N1)C(=O)OCC1=C(C)OC(=O)O1)C(C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Olmesartan medoxomil
Summenformel C29H30N6O6
Menge 1 g
Molekulargewicht (g/mol) 558.60
Formelmasse 558.59

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'