Learn More
Euro Protection™ Okenite Safety Shoes
missing translation for 'orderingAttributeHoverText'
Größe:
37
38
39
40
41
42
43
44
45
46
47
missing translation for 'unitSize'
1 Stück
Description
- Suitable for food industry use
- Easy-on and easy-off
- Protective cover
- Rounded heel for more comfortable driving
- Microfiber upper
- Breathable 3D mesh lining
- Removable, textile and foam insole
- Composite toe cap
- Injected polyurethane midsole
- Polyurethane outsole
Specifications
Specifications
| Farbe | White, Gray |
| Zur Verwendung mit (Anwendung) | Suitable for food industry |
| Fersenform | Rounded |
| Auskleidungsmaterial | Atmungsaktives 3D-Mesh-Futter |
| Material | Microfiber |
| Größe | 46 |
| Sohleneigenschaften | Removable, textile and foam insole, injected polyurethane midsole, polyurethane outsole. |
| Sohlenmaterial | Polyurethan |
| Modell | Low-cut |
| Zehenmaterial | Komposit |
| Show More |