Learn More
Brady™ Nylon Printer Labels for ID Pro Plus, LS 2000 and BradyMarker XC Plus
Blank, printable, nylon cloth labels for the ID Pro™ Plus, LS 2000™ and BradyMarker printers.
Marke: Brady™ 232624
Additional Details : Gewicht : 0.03400kg
Beschreibung
This material is a UL-Recognised component when printed with R2000 and R5000 dot matrix ribbons.
- High adhesion
- Low profile
Spezifikation
Autoclavable | |
Yellow | |
Nylon Cloth (B-499) | |
Non-sterile | |
I.D. PRO Handheld Labeler, LS2000 Labeling System | |
500 Labels | |
-40°C to +90°C | |
12.7 mm | |
7.1 mm |
Permanent Acrylic | |
Matte | |
Thermal Transfer | |
Equipment and Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking | |
1 | |
Rectangle | |
0.17 mm | |
12.7 mm | |
7 mm |