Learn More
Invitrogen™ NuPAGE™ WedgeWell™ Midi Tris-Acetate Welcome Pack, 3 to 8%
Beschreibung
The NuPAGE Tris-Acetate Midi Welcome Pack, Wedgewell format provides all the necessary NuPAGE gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank.
The NuPAGE Tris-Acetate Midi Wedgewell Welcome Pack provides all the necessary NuPAGE gels, buffers, and reagents needed to begin using the SureLock Tandem Midi Gel Tank.
Welcome Pack contents
- SureLock Tandem Midi Gel Tank (STM1001)
- NuPAGE Tris-Acetate WedgeWell Midi Gels (2 boxes, 20 gels)
- NuPAGE Tris-Acetate SDS Running Buffer, 20X (LA0041)
- NuPAGE LDS Sample Buffer, 4X (NP0007)
- NuPAGE Sample Reducing Agent, 10X (NP0009)
- HiMark Pre-stained Protein Standard (LC5699)
About the SureLock Tandem Midi Gel Tank
The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a setup time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.
About NuPAGE Tris-Acetate Midi gels
NuPAGE Tris-Acetate Midi protein gels provide excellent separation of large molecular weight proteins. NuPAGE Tris-Acetate Midi gels are high performance polyacrylamide gels that simulate the denaturing or the native conditions of the traditional Laemmli system. The unique buffer formulation maintains a low operating pH during electrophoresis, resulting in superior resolution of proteins compared to traditional Tris-glycine midi SDS-PAGE gels.
Transfer and detection
For transfer of proteins to a membrane, we recommend using NuPAGE Transfer Buffer (NP0006) for traditional wet transfer using the SureLock Tandem Midi Blot Module (STM2001). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 3 Gel Transfer Device (IB31001).
Convenient to use
NuPAGE WedgeWell-format Tris-Acetate gels have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0-mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.
Spezifikation
Spezifikation
| Zur Verwendung mit (Geräte) | SureLock Tandem Midi-Gel-Tank |
| Gelgröße | Midi |
| Produktlinie | NuPAGE |
| Gelanteil (%) | 3 to 8% |
| Geltyp | Tris-Acetate |
| Anzahl Wells | 20-Well |
| Menge | 1 Welcome Pk. |
| Trennungsbereich | 40 to 500 kDa |
| Versandbedingung | Zugelassen für den Versand bei Raumtemperatur und auf Nasseis |
| Dicke (metrisch) | 1.0 mm |
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.