missing translation for 'onlineSavingsMsg'
Learn More

NUP53 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 19819-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839304

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The elongation of primed DNA templates by DNA polymerase delta and DNA polymerase epsilon requires the accessory proteins proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also named activator 1, is a protein complex consisting of five distinct subunits of 140, 40, 38, 37, and 36 kDa. This gene encodes the 38 kDa subunit. This subunit is essential for the interaction between the 140 kDa subunit and the core complex that consists of the 36, 37, and 40 kDa subunits. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Spezifikation

Spezifikation

NUP53
Polyclonal
Unconjugated
NUP35
35 kDa nucleoporin, Mitotic phosphoprotein 44, MP 44, MP44, nucleoporin 35kDa, Nucleoporin Nup35, Nucleoporin NUP53, NUP35, NUP53
Rabbit
Antigen Affinity Chromatography
RUO
129401, 295692, 69482
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q68FY1, Q8NFH5, Q8R4R6
Nup35
NUP53 Fusion Protein Ag13869
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote