missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP50 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20798-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. NUP50 is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. NUP50 may serve as a binding site on the nuclear side of the pore complex for export receptor-cargo complexes. It interacts with multiple transport receptor proteins including p27Kip1. This interaction is required for correct intracellular transport and degradation of p27Kip1.Spezifikation
NUP50 | |
Polyclonal | |
Unconjugated | |
NUP50 | |
50 kDa nucleoporin, nucleoporin 50kDa, Nucleoporin Nup50, NUP50 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10762, 18141, 25497 | |
Antibody | |
IgG |
Western Blot | |
0.57 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08587, Q9JIH2, Q9UKX7 | |
Nup50 | |
NUP50 Fusion Protein Ag14710 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |