Learn More
NUDT8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559580
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KCDPADQDVV HTALRETREE LGLAVPEEHV WGLLRPVYDP QKATVVPVLA GVGPLDPQSL RPNSEEVD Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
Probably mediates the hydrolysis of some nucleoside diphosphate derivatives.Spezifikation
NUDT8 | |
Polyclonal | |
Unconjugated | |
Nudt8 | |
2310039H17Rik; Nucleoside diphosphate-linked moiety X motif 8; nucleoside diphosphate-linked moiety X motif 8, mitochondrial; nudix (nucleoside diphosphate linked moiety X)-type motif 8; nudix hydrolase 8; nudix motif 8; nudix-type motif 8; Nudt8 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254552 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WV74 | |
Nudt8 | |
Recombinant protein corresponding to Human NUDT8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only