Learn More
Nuclear Membrane Marker Antibody (AE-5), Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-11307-20ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Spezifikation
| Nuclear Membrane Marker | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunocytochemistry/ Immunofluorescence 1-2 μg/ml | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Immunofluorescence | |
| AE-5 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Nuclei of myeloid leukemia biopsy cells | |
| 20 μg | |
| Primary | |
| Store at 4°C. | |
| IgG1 κ |