Learn More
Invitrogen™ Nuclear Matrix Protein p84 Recombinant Rabbit Monoclonal Antibody (24GB2205)
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA557773
 Lernen Sie weitere Varianten dieses
Artikelnummer. 30274872
  • 190.00 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Nuclear Matrix Protein p84 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Nuclear Matrix Protein p84
Recombinant Monoclonal
Unconjugated
Thoc1
3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1
Rabbit
Affinity Chromatography
RUO
225160, 291797, 9984
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2205
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P59924, Q8R3N6, Q96FV9
Thoc1
A synthesized peptide derived from human Nuclear Matrix Protein p84 (aa 350-400).
20 μL
Primary
Human, Mouse, Rat