Learn More
NRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10724-1-AP-150UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16847293
  • 385.92 CHF / 150 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noo syndrome 6.
Spezifikation
Spezifikation
NRAS
Polyclonal
Unconjugated
NRAS
ALPS4, GTPase NRas, HRAS1, N ras, NRAS, NRAS1, Transforming protein N Ras
Rabbit
Antigen Affinity Chromatography
RUO
18176, 24605, 4893
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01111, P08556, Q04970
Nras
NRAS Fusion Protein Ag1081
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen