missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPLOC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11638-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
NPLOC4 gene ontology annotations related to this gene include ubiquitin-dependent protein catabolic process.Spezifikation
NPLOC4 | |
Polyclonal | |
Unconjugated | |
NPLOC4 | |
KIAA1499, NPLOC4, Protein NPL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140639, 217365, 55666 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60670, Q8TAT6, Q9ES54 | |
NPLOC4 | |
NPLOC4 Fusion Protein Ag2219 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |