missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPFFR2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19505-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody can recognize isoform 1, isoform 2, and isoform 3 of NPFFR2.
Receptor for NPAF (A-18-F-amide) and NPFF (F-8-F-amide) neuropeptides, also known as morphine-modulating peptides. Can also be activated by a variety of naturally occurring or synthetic FMRF-amide like ligands. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.Spezifikation
NPFFR2 | |
Polyclonal | |
Unconjugated | |
NPFFR2 | |
G protein coupled receptor 74, GPR74, HLWAR77, neuropeptide FF receptor 2, NPFFR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10886 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y5X5 | |
NPFFR2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |