missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTCH1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | NOTCH1 |
---|---|
Konzentration | 0.3 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850265
|
Proteintech
20687-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840265
|
Proteintech
20687-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 09-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The antibody is specific to NOTCH1. It can recognize the full length NOTCH1 (270 kDa) and all the three cleaved NOTCH1 forms.
This gene is similar in sequence to the S. cerevisiae YDL201w gene. The gene product contains a conserved S-adenosylmethionine-binding motif and is inactivated by phosphorylation.Spezifikation
NOTCH1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P46531 | |
4851 | |
Peptide | |
Primary | |
-20°C | |
NOTCH1 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hN1, Notch 1, NOTCH1, TAN1 | |
NOTCH1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |