Learn More
Noradrenalone Hydrochloride, TRC
Artikelnummer. 30007629
Change view
Click to view available options
Menge:
100 mg
500 mg
1 g
Packungsgröße:
1 Gramm
100 Milligramm
500 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30007629 1 g 1 Gramm
30011992 500 mg 500 Milligramm
17918104 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007629 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCN6610351G
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 5090-29-9
Summenformel C8H10ClNO3
Molekulargewicht (g/mol) 203.62
Synonym 2-Amino-3',4'-dihydroxyacetophenone Hydrochloride, 2-Amino-1-(3,4-dihydroxyphenyl)ethanone Hydrochloride
IUPAC-Name 2-amino-1-(3,4-dihydroxyphenyl)ethanone;hydrochloride
SMILES Cl.O=C(C1=CC=C(O)C(O)=C1)CN
Spezifikation
Chemischer Name oder Material Noradrenalone Hydrochloride
Menge 1 g
InChI-Formel InChI=1S/C8H9NO3.ClH/c9-4-8(12)5-1-2-6(10)7(11)3-5;/h1-3,10-11H,4,9H2;1H
Formelmasse 203.0349