Learn More
NMDA R, NR1 Subunit Antibody - Prime Splice Variant - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB300-117
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NMDA R, NR1 Subunit Polyclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Spezifikation
| NMDA R, NR1 Subunit | |
| Polyclonal | |
| Unconjugated | |
| P35439 | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Reconstitute with 50 uL PBS to desired concentration. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| LYOPH | |
| Western Blot 1:1000, Immunohistochemistry 1:1000 - 1:2000, Immunohistochemistry-Frozen 1:1000 - 1:2000 | |
| GluN1, MRD8, NDHMSD, NDHMSR;NMD-R1, NMDA1, NMDAR1, NR1 | |
| Synthetic peptide corresponding to amino acid residues specific to the NR1 subunit, C2' splice variant insert conjugated to KLH. Accession # P35439 | |
| 0.025 mg | |
| Cellular Signaling, Neuroscience | |
| 2902 | |
| Mouse, Rat | |
| Purified |