missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Rabbit, Clone: NX2/1422R, enQuireBio™
Rabbit Monoclonal Antibody
Marke: EnquireBio 4821-RBM2-P0
Additional Details : Gewicht : 0.01000kg
Spezifikation
NKX2.2 | |
Nuclear | |
NX2/1422R | |
Unconjugated | |
XP_006723629 | |
Rabbit | |
40-50kDa | |
Protein A/G Purified | |
RUO | |
4821 | |
IgG κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
NKX2-2 | |
Recombinant human NKX2.2 protein fragment | |
Pancreas or Ewing s sarcoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |