missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK-1.1 Mouse anti-Mouse, BB700, Clone: PK136, BD Horizon™
Mouse Monoclonal Antibody
Marke: BD Biosciences 566502
Additional Details : Gewicht : 0.00840kg
Beschreibung
NK-1.1 Monoclonal antibody specifically detects NK-1.1 in Mouse samples. It is validated for Flow Cytometry.Spezifikation
NK-1.1 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
PK136 | |
Brilliant Blue 700 (BB700) | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |