missing translation for 'onlineSavingsMsg'
Learn More

Nicotinsäure, 99 %, Thermo Scientific Chemicals

Catalog Number p-7020650
Click to view available options
Menge:
250 g
1000 g
5000 g
59-67-6
C6H5NO2
123.11
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
938
CHEBI:15940
Pyridin-3-Carbonsäure
OC(=O)C1=CC=CN=C1
59-67-6
C6H5NO2
123.11
MFCD00006391
PVNIIMVLHYAWGP-UHFFFAOYSA-N
nicotinic acid, niacin, 3-pyridinecarboxylic acid, 3-carboxypyridine, wampocap, acidum nicotinicum, apelagrin, pellagrin, akotin, daskil
938
CHEBI:15940
Pyridin-3-Carbonsäure
OC(=O)C1=CC=CN=C1

Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Wasserlöslich. Schwer löslich in Ethanol und Ethylether. Unlöslich in den meisten Lipidlösungsmitteln.

Hinweise
Lichtempfindlich. Kühl lagern. Nicht kompatibel mit starken Oxidationsmitteln.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Nicotinic Acid
Schmelzpunkt 236°C to 239°C
Dichte 1.4
Flammpunkt 293°C (559°F)
Prozentgehaltsbereich 99%
Menge 250 g
Beilstein 109591
Merck Index 14,6525
Löslichkeitsinformationen Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.
Formelmasse 123.11
Reinheit (%) 99%
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'