missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE3/SLC9A3, p Ser552 Antibody (14D5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NB110-81529H
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Knockout Validated, Knockdown Validated.
Spezifikation
| NHE3/SLC9A3 (p Ser552) | |
| Monoclonal | |
| HRP | |
| isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3 | |
| Mouse | |
| 85 kDa | |
| 0.1 mL | |
| Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| 6550 | |
| Human, Mouse, Rat, Pig, Opossum, Primate, Rabbit | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoassay, KnockDown | |
| 14D5 | |
| Western Blot, Simple Western, ELISA, Immunohistochemistry, Knockout Validated, Knockdown Validated | |
| SLC9A3 | |
| Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433] | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody detects NHE3 posphorylated at Serine 552, and does not cross-react with the non-phosphopeptide. | |
| Store at 4C in the dark. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur