Learn More
NHE3/SLC9A3, p Ser552 Antibody (14D5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 15 publications
Marke:  Novus Biologicals NB110-81529
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18791494
  • 568.00 CHF / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
NHE3/SLC9A3 Monoclonal specifically detects NHE3/SLC9A3 in Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
NHE3/SLC9A3 (p Ser552)
Monoclonal
2 mg/mL
Western Blot 1-5 ug/ml, Simple Western 1:1000, Flow Cytometry reported in scientific literature (PMID 21255452), ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Knockout Validated reported in scientific literature (PMID 31276916), KnockDown Validated reported in scientific literature (PMID 31276916)
SLC9A3
Synthetic peptide made to a region surrounding the Serine 552 portion of rat NHE3. [UniProt# P26433]
Protein G purified
RUO
Primary
This antibody detects NHE3 posphorylated at Serine 552, and does not cross-react with the non-phosphopeptide.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay, KnockDown
14D5
Unconjugated
isoform 3, MGC126718, NHE3MGC126720, solute carrier family 9 (sodium/hydrogen exchanger), member 3, Solute carrier family 9 member 3
Mouse
85 kDa
0.1 mL
Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
6550
Human, Mouse, Rat, Porcine, Opossum, Primate, Rabbit
Purified
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen