missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nhe-1 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15975595
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
15975595 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15975595 Lieferant Invitrogen™ Lieferanten-Nr. PA580033

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat PC-12 whole cell, human MCF-7 whole cell. IHC: rat kidney tissue, human mammary cancer tissue, mouse kidney tissue.

The Na+/H+ antiporter (Nhe-1) is a ubiquitous membrane-bound enzyme involved in pH regulation of vertebrate cells and is specifically inhibited by the diuretic drug amiloride and activated by a variety of signals including growth factors, mitogens, neurotransmitters, and tumor promoters. Nhe-1 acts as an anchor for actin filaments to control the integrity of the cortical cytoskeleton. This occurs through a previously unrecognized structural link between Nhe-1 and the actin-binding proteins ezrin, radixin, and moesin, collectively referred to as ERM proteins. A structural role for Nhe-1 has been proposed in regulating the cortical cytoskeleton that is independent of its function as an ion exchanger. It is also thought that Nhe-1 play a role in hypertension. At least two isoforms of Nhe-1 are known to exist.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Nhe-1
Anwendungen Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 500 μg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 5mg BSA and 0.05mg sodium azide
Gen SLC9A1
Gen-Zugriffsnummer P19634, P26431, Q61165
Gen-Alias Apnh; APNH1; AW554487; LIKNS; Mir5122; mir-5122; Na(+)/H(+) antiporter, amiloride-sensitive; Na(+)/H(+) exchanger 1; Na+/H+ antiporter; Na+/H+, amiloride sensitive; Na-Li countertransporter; Nhe1; NHE-1; PPP1R143; protein phosphatase 1, regulatory subunit 143; SLC9A1; slow-wave epilepsy; sodium/hydrogen exchanger 1; Solute carrier family 9 (sodium/hydrogen exchanger 1), antiporter, Na+/H+, (amiloride sensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 1 (antiporter, Na+/H+, amiloride sensitive); solute carrier family 9 (sodium/hydrogen exchanger), member 1; solute carrier family 9 (sodium/hydrogen exchanger), member 1 (antiporter, Na+/H+, amiloride sensitive); solute carrier family 9 member 1; solute carrier family 9 member A1; solute carrier family 9, member 1; solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1; swe
Gensymbole SLC9A1
Wirtsspezies Rabbit
Immunogen E.coli-derived human SLC9A1 recombinant protein (Position: H543-Q815).
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 20544, 24782, 6548
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Lyophilized
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.