Beschreibung
- Stable solution at room temperature with neutral pH, and also maintains cell integrity
- Recovered cells can be easily sub-cultured in both 2D and 3D formats
- The solution is versatile and can be used before or after the fixation and staining of hydrogel specimens, ensuring high-quality data for downstream analysis
- It is suitable for harvesting cells from 2D ECM-coated plates
- Ensures a high recovery rate and cell viability, making it perfect for passaging, cryopreservation, or further biochemical analysis
Spezifikation
Spezifikation
| Form | Liquid |
| Produkttyp | Organoid Harvesting Solution |
| Sterilität | Sterile |
| Sterilization Method | Autoclaved |
| Menge | 500 mL |