missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neutrophil elastase Monoclonal Antibody (ELA11-207.2), Invitrogen™

Mouse Monoclonal Antibody
720.00 CHF
Spezifikation
| Antigen | Neutrophil elastase |
|---|---|
| Klon | ELA11-207.2 |
| Konzentration | 1.8 mg/mL |
| Anwendungen | ELISA |
| Klassifikation | Monoclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
11568892
![]() |
Invitrogen™
MA110606 |
500 μg |
720.00 CHF
500 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
MA1-10606 detects Neutrophil elastase from human samples. MA1-10606 has been successfully used in ELISA applications. The MA1-10606 immunogen is purified human Neutrophil Elastase.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.Spezifikation
| Neutrophil elastase | |
| 1.8 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| ELANE | |
| IgM | |
| Protein A | |
| Antibody |
| ELA11-207.2 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08246 | |
| 1991 | |
| Purified human Neutrophil Elastase. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ELANE |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts