missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-1 (CD304) Mouse anti-Human, APC-R700, Clone: U21-1283, BD Horizon™
Mouse Monoclonal Antibody
311.00 CHF - 690.00 CHF
Spezifikation
Antigen | Neuropilin-1 (CD304) |
---|---|
Klon | U21-1283 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | APC-R700 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15824709
|
BD Biosciences
566039 |
25 Tests |
311.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15814709
|
BD Biosciences
566038 |
100 Tests |
690.00 CHF
1 Stück |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.
Neuropilin-1 (CD304) Monoclonal antibody specifically detects Neuropilin-1 (CD304) in Human samples. It is validated for Flow cytometry.Spezifikation
Neuropilin-1 (CD304) | |
Flow Cytometry | |
APC-R700 | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
Recombinant Human Neuropilin-1 | |
Primary | |
Vol. Per test: 5μL |
U21-1283 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Neuropilin; NRP; NP1; Blood dendritic cell antigen 4; BDCA-4; VEGF165R | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. |
For Research Use Only.