Learn More
Netilmicin sulfate, 595 ^mg/mg, Thermo Scientific™
A broad spectrum antibiotic
154.33 CHF - 270.31 CHF
Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15427609
Sicherheitsdatenblatt (SDS) Unterlagen ansehen
Thermo Scientific Alfa Aesar
J66302.MA
154.33 CHF
10 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Lager vorrätig
Zum Warenkorb hinzufügen
 Eingeschränkter Artikel
15437609
Sicherheitsdatenblatt (SDS) Unterlagen ansehen
Thermo Scientific Alfa Aesar
J66302.MB
25mg
270.31 CHF
25 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 31-08-2026
Zum Warenkorb hinzufügen
 Eingeschränkter Artikel
Beschreibung
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Netilmicin sulfate is used as a broad spectrum antibiotic active against most Gram-negative and some Gram-positive bacteria. It is a semisynthetic 1-N-ethyl derivative of sisomycin, an aminoglycoside antibiotic with action similar to gentamicin, but has less ear and kidney toxicity. It is used in aminoglycoside resistance studied
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.