Learn More
Nestin Monoclonal Antibody (10C2), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
283.00 CHF - 600.00 CHF
Spezifikation
Antigen | Nestin |
---|---|
Klon | 10C2 |
Konzentration | 0.5 mg/mL |
Anwendungen | Flow Cytometry, Immunocytochemistry, Immunohistochemistry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15302090
|
Affymetrix eBioscience
53-9843-80 |
25 μg |
283.00 CHF
25 Mikrogramm |
Verfügbar ab: 29-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15312090
|
Affymetrix eBioscience
53-9843-82 |
100 μg |
600.00 CHF
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Description: The monoclonal antibody 10C2 recognizes human nestin (residues 1464-1614). Nestin is a 220-240 kDa Class VI intermediate filament protein that is expressed in stem cells of the developing nervous system. Expression has also been found in pancreatic islet and mesenchymal precursors in addition to cells with angiogenic potential. Upon differentiation, nestin-containing filaments are replaced by cell type-specific intermediate filament proteins, such as GFAP. Expression has been identified in a variety of cancerous cells including glioblastomas and pancreatic tumors. No cross-reactivity to rodent nestins has been observed using the 10C2 antibody. Applications Reported: This 10C2 antibody has been reported for use in microscopy, immunocytochemistry, immunohistochemical staining, and flow cytometric analysis. Applications Tested: This 10C2 antibody has been tested by immunocytochemistry of formaldehyde-fixed and permeabilized cells. This can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Spezifikation
Nestin | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AA166324; C78523; ESTM46; FLJ21841; intermediate filament protein; Nbla00170; Nes; Nestin | |
Nes | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Nes |
10C2 | |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
P48681 | |
10763 | |
IgG1 | |
Affinity chromatography | |
Antibody |
For Research Use Only.