missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NEK6, GST Tag Recombinant Protein

Artikelnummer. 17178705
Change view
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17178705 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17178705 Lieferant Invitrogen™ Lieferanten-Nr. A31519

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with our Z'-LYTE™ assay technology to identify novel kinase inhibitors. NEK6 is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE assay technology Available in multiple pack sizes for convenient use Also available in our SelectSceen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for NEK6 is NP_055212.2.

NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition. The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA. NEK6 encodes a NEK type protein kinase, which contains a C-terminal NIMA-like catalytic domain.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q9HC98
Konzentration See Label
Zur Verwendung mit (Anwendung) Kinase Assay
Zusammensetzung 50mM tris HCl with 10mM glutathione, 150mM NaCl, 0.1mM PMSF, 0.1mM EDTA, 0.25mM DTT, 25% glycerol and no preservative; pH 7.5
Gen-ID (Entrez) 10783
Molekulargewicht 62.4 kDa
Name Human NEK6, GST Tag
pH-Bereich 7.5
Reinigungsverfahren Purified
Menge 1 mg
Lagerungsbedingungen -80°C, Avoid Freeze/Thaw Cycles
Kennzeichnung RUO
Gen-Alias 1300007C09Rik; Nek6; Never in mitosis A-related kinase 6; NIMA (never in mitosis gene a)-related expressed kinase 6; NIMA (never in mitosis gene a)-related kinase 6; NIMA related kinase 6; NIMA-related expressed kinase 6; NIMA-related kinase 6; nimA-related protein kinase 6; Protein kinase SID6-1512; putative serine-threonine protein kinase; RP11-101K10.6; serine/threonine-protein kinase Nek6; SID; SID6-1512
Gebräuchliche Bezeichnung NEK6
Gensymbol NEK6
Kinase-Familie NEKs
Proteinlänge Full Length
Produkttyp Protein
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung GST-tag
Sequenz Full length
Inhalt und Lagerung Storage Buffer: 50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1mM PMSF, and 25% Glycerol.

Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws.
Versandbedingung Dry Ice
Expressionssystem Insect cells
Proteinform Full Length, Recombinant, Active
Form Liquid
Forschungskategorie Drug Development
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.