missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK6 Recombinant Human Protein
NEK6 kinase is a member of the NEK family. NEK6 is a serine/threonine kinase that controls the initiation of mitosis and is required for the metaphase-anaphase transition.
850.00 CHF - 2605.00 CHF
Spezifikation
Zugriffsnummer | NP_055212.2 |
---|---|
Zur Verwendung mit (Anwendung) | Kinase Assay, Target and Lead Identification and Validation, Kinase Biology, Kinase Assay, Industrial and Applied Science, Pharma and Biopharma |
Zusammensetzung | 50mM Tris-HCl (pH 7.5), 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, and 25% Glycerol. |
Molekulargewicht | 62.4kDa |
Lagerungsbedingungen | Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15607888
|
Invitrogen™
A31511 |
10μg |
850.00 CHF
10 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15617888
|
Invitrogen™
A31512 |
100μg |
2605.00 CHF
100 Mikrogramm |
Verfügbar ab: 30-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with our Z'-LYTE™ assay technology to identify novel kinase inhibitors. NEK6 is also available in our SelectScreen™ Kinase Profiling Service.
- Validated for use with the Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Spezifikation
NP_055212.2 | |
50mM Tris-HCl (pH 7.5), 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, and 25% Glycerol. | |
Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws. | |
SID6-1512 | |
Full Length | |
Recombinant | |
Insect |
Kinase Assay, Target and Lead Identification and Validation, Kinase Biology, Kinase Assay, Industrial and Applied Science, Pharma and Biopharma | |
62.4kDa | |
Full Length, Recombinant, Active | |
NEKs | |
Human | |
GST Tag |