missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12015-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
NDRG2′s gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Spezifikation
NDRG2 | |
Polyclonal | |
Unconjugated | |
NDRG2 | |
KIAA1248, NDRG family member 2, NDRG2, Protein NDRG2, Protein Syld709613, SYLD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171114, 29811, 57447 | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VBU2, Q9QYG0, Q9UN36 | |
Ndrg2 | |
NDRG2 Fusion Protein Ag2629 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |