Learn More
NCOA2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 30962-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The NCOA2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NCOA2. This antibody recognizes human antigen. The NCOA2 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| NCOA2 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| bHLHe75, GRIP1, hTIF2, KAT13C, NCoA 2, NCOA2, nuclear receptor coactivator 2, SRC2, TIF2 | |
| 20 μL | |
| 10499 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |