Learn More
Naltrexone hydrochloride, Thermo Scientific™
A congener of naloxone and also an opioid antagonist
157.18 CHF - 461.24 CHF
Chemische Identifikatoren
| CAS | 16676-29-2 |
|---|---|
| Summenformel | C20H24ClNO4 |
| Molekulargewicht (g/mol) | 377.865 |
| MDL-Nummer | MFCD00069324 |
| InChI-Schlüssel | RHBRMCOKKKZVRY-ITLPAZOVSA-N |
| Synonym | naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin |
| PubChem CID | 5485201 |
| IUPAC-Name | (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride |
| SMILES | C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15463339
|
Thermo Scientific Alfa Aesar
J60590.MD |
250mg |
171.00 CHF 157.18 CHF / 250 Milligramm Sparen 13.82 CHF 8% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15453339
|
Thermo Scientific Alfa Aesar
J60590.03 |
1g |
498.00 CHF 461.24 CHF / 1 Gramm Sparen 36.76 CHF 7% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsA congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.
Solubility
Soluble in water at 50mg/ml.
Notes
Light sensitive. Incompatible with bases and oxidizing agents. Store in a cool place.
Chemische Identifikatoren
| 16676-29-2 | |
| 377.865 | |
| RHBRMCOKKKZVRY-ITLPAZOVSA-N | |
| 5485201 | |
| C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl |
| C20H24ClNO4 | |
| MFCD00069324 | |
| naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin | |
| (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride |
Spezifikation
| Naltrexone hydrochloride | |
| C20H24ClNO4 | |
| 3580333 | |
| 14,6363 | |
| Soluble in water at 50mg/ml. | |
| C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl | |
| 377.865 | |
| 377.86 |
| 16676-29-2 | |
| MFCD00069324 | |
| Light sensitive | |
| naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin | |
| RHBRMCOKKKZVRY-ITLPAZOVSA-N | |
| (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride | |
| 5485201 |
Sicherheit und Handhabung
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 240-723-0
missing translation for 'rtecsNumber' : QD2160000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.