missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-Nomega-(2,2,4,6,7-Pentamethyl-2,3-Dihydrobenzo[b]furan-5-ylsulfonyl)-D-Arginin, 95 %, Thermo Scientific Chemicals

Catalog Number 15435585
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
186698-61-3
C24H38N4O7S
526.649
MFCD00236822
CVFXPOKENLGCID-QGZVFWFLSA-N
boc-d-arg pbf-oh, boc-nw-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine, ambotzbaa1021, 2r-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2r-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid, e-n∼5∼-amino 2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonyl amino methylidene-n∼2∼-tert-butoxycarbonyl-d-ornithine, n-alpha-t-butyloxycarbonyl-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine
16213160
(2R)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methyliden]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentansäure
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
186698-61-3
C24H38N4O7S
526.649
MFCD00236822
CVFXPOKENLGCID-QGZVFWFLSA-N
boc-d-arg pbf-oh, boc-nw-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine, ambotzbaa1021, 2r-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2r-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid, e-n∼5∼-amino 2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-sulfonyl amino methylidene-n∼2∼-tert-butoxycarbonyl-d-ornithine, n-alpha-t-butyloxycarbonyl-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine
16213160
(2R)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methyliden]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentansäure
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine
Formelmasse 526.65
Reinheit (%) 95%
Menge 250 mg
Physikalische Form Fest

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'