missing translation for 'onlineSavingsMsg'
Learn More

N-Methyldesloratadin, TRC

Product Code. 17904415
Change view
Click to view available options
Menge:
250 mg
Unit Size:
250 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17904415 250 mg 250 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17904415 Supplier Toronto Research Chemicals Supplier No. TRCM316500250MG
 Eingeschränkter Artikel
Verfügbar ab: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38092-89-6
Summenformel C20 H21 Cl N2
Molekulargewicht (g/mol) 324.85
Synonym 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Loratadine Imp. G (EP)
IUPAC-Name 8-Chloro-11-(1-Methylpiperidin-4-Yliden)-5,6-Dihydrobenzo[1,2]Cyclohepta[2,4-b]Pyridin
SMILES CN1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1

Specifications

Chemischer Name oder Material 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Hinweise zur Reinheitsqualität HPLC
Menge 250 mg
InChI-Formel InChI=1S/C20H21ClN2/c1-23-11-8-14(9-12-23)19-18-7-6-17(21)13-16(18)5-4-15-3-2-10-22-20(15)19/h2-3,6-7,10,13H,4-5,8-9,11-12H2,1H3
Formelmasse 324.1393
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.