Learn More
N-Lauroylsarcosin-Natriumsalz, MP Biomedicals™
Artikelnummer. 11456471 Alle ansehen MP Biomedicals Produkte
Change view
Click to view available options
Menge:
50 g
100 g
250 g
Packungsgröße:
250 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11456471 100 g -
11446471 50 g -
11466471 250 g 250 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11456471 Lieferant MP Biomedicals Lieferanten-Nr. 0219400880
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Chemische Identifikatoren
CAS 137-16-6
Summenformel C15H28NNaO3
Molekulargewicht (g/mol) 293.38
MDL-Nummer MFCD00042728
InChI-Schlüssel KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
Spezifikation
Chemischer Name oder Material N-Lauroylsarcosine Sodium Salt
Namenshinweis Molecular biology reagent
Farbe Weiß
Prozentgehaltsbereich >95.5%
Menge 100 g
IUPAC-Name Natrium 2-(N-Methyldodecanamido)acetat
Formelmasse 293.4
Physikalische Form Pulver