missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Antibody (13A9), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 35 publications
271.00 CHF - 631.00 CHF
Spezifikation
| Antigen | N-Cadherin |
|---|---|
| Klon | 13A9 |
| Konzentration | 1 mg/mL |
| Verdünnung | Western Blot 0.5 ug/ml, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Flow (Intracellular), Immunocytochemistry |
| Anwendungen | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18740224
|
Novus Biologicals
NBP1-48309SS |
0.025 mL |
271.00 CHF
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18758303
|
Novus Biologicals
NBP1-48309 |
0.1 mL |
631.00 CHF
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
N-Cadherin Monoclonal specifically detects N-Cadherin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunocytochemistry.Spezifikation
| N-Cadherin | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction, Stem Cells | |
| P19022 | |
| 1000 | |
| This N-Cadherin Antibody (13A9) was developed against the cytoplasmic domain of human N Cadherin [Swiss-Prot# P19022]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 13A9 | |
| Western Blot 0.5 ug/ml, Simple Western 1:50, Flow Cytometry, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Flow (Intracellular), Immunocytochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cadherin 2, N-cadherin (neuronal), cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, CD325, CD325 antigen, CDHNcalcium-dependent adhesion protein, neuronal, CDw325, N-cadherin, NCADN-cadherin 1, Neural cadherin, neural-cadherin | |
| CDH2 | |
| IgG1 | |
| Protein G purified | |
| 140 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts