Learn More
N-Boc-L-Valinyl-L-Leucinylanilid, TRC
Specifications
Specifications
| Chemischer Name oder Material | N-Boc-L-valinyl-L-leucinyl Anilide |
| CAS | 874899-05-5 |
| Summenformel | C22 H35 N3 O4 |
| Menge | 5 mg |
| InChI-Formel | InChI=1S/C22H35N3O4/c1-14(2)13-17(19(26)23-16-11-9-8-10-12-16)24-20(27)18(15(3)4)25-21(28)29-22(5,6)7/h8-12,14-15,17-18H,13H2,1-7H3,(H,23,26)(H,24,27)(H,25,28)/t17-,18-/m0/s1 |
| SMILES | CC(C)C[C@H](NC(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C)C(=O)Nc1ccccc1 |
| IUPAC-Name | tert-Butyl-N-[(2S)-1-[[(2S)-1-Anilino-4-methyl-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]carbamat |
| Molekulargewicht (g/mol) | 405.53 |
| Formelmasse | 405.2628 |