missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-Methionin, 98+ %, Thermo Scientific Chemicals

Catalog Number p-7045040
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5241-66-7
C10H19NO4S
249.33
MFCD00038256
IMUSLIHRIYOHEV-SSDOTTSWSA-N
boc-d-met-oh, boc-d-methionine, n-boc-d-methionine, n-tert-butoxycarbonyl-d-methionine, n-tert-butyloxycarbonyl-d-methionine, boc-l-met-oh, d-methionine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-methionine, n-alpha-t-butyloxycarbonyl-d-methionine
111051
CSCC[C@@H](NC(=O)OC(C)(C)C)C(O)=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
5241-66-7
C10H19NO4S
249.33
MFCD00038256
IMUSLIHRIYOHEV-SSDOTTSWSA-N
boc-d-met-oh, boc-d-methionine, n-boc-d-methionine, n-tert-butoxycarbonyl-d-methionine, n-tert-butyloxycarbonyl-d-methionine, boc-l-met-oh, d-methionine, n-1,1-dimethylethoxy carbonyl, tert-butoxycarbonyl-d-methionine, n-alpha-t-butyloxycarbonyl-d-methionine
111051
CSCC[C@@H](NC(=O)OC(C)(C)C)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Boc-D-methionine
Schmelzpunkt 47°C to 50°C
Menge 1 g
Beilstein 4294122
Optische Drehung +22° (c=1 in Methanol)
IUPAC-Name (2R)-2-{[(tert-Butoxy)carbonyl]amino}-4-(methylsulfanyl)butanoat
Formelmasse 249.32
Reinheit (%) ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'