Learn More
N-Benzyloxycarbonylglycin, 98 +%, Thermo Scientific Chemicals
Catalog Number p-7022935
Click to view available options
Menge:
25 g
100 g
500 g
1138-80-3
C10H11NO4
209.201
MFCD00002691
CJUMAFVKTCBCJK-UHFFFAOYSA-N
n-carbobenzyloxyglycine, n-cbz-glycine, z-gly-oh, carbobenzyloxyglycine, carbobenzoxyglycine, n-benzyloxycarbonylglycine, n-carbobenzoxyglycine, cbz-gly-oh, cbz gly, benzyloxycarbonylglycine
14349
CHEBI:16532
2-(Phenylmethoxycarbonylamino)essigsäure
C1=CC=C(C=C1)COC(=O)NCC(=O)O
1138-80-3
C10H11NO4
209.201
MFCD00002691
CJUMAFVKTCBCJK-UHFFFAOYSA-N
n-carbobenzyloxyglycine, n-cbz-glycine, z-gly-oh, carbobenzyloxyglycine, carbobenzoxyglycine, n-benzyloxycarbonylglycine, n-carbobenzoxyglycine, cbz-gly-oh, cbz gly, benzyloxycarbonylglycine
14349
CHEBI:16532
2-(Phenylmethoxycarbonylamino)essigsäure
C1=CC=C(C=C1)COC(=O)NCC(=O)O
N-Carbobenzoxyglycine is used in the dipeptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
N-Carbobenzoxyglycin wird bei der Dipeptidsynthese verwendet.
Löslichkeit
Löslich in Methanol. Nicht in Wasser löslich.
Hinweise
An einem kühlen und trockenen Ort lagern. Von Hitze und Oxidationsmitteln fernhalten.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Benzyloxycarbonylglycine
Schmelzpunkt 119°C to 122°C
Strukturformel C6H5CH2OCONHCH2CO2H
Beilstein 526877
Löslichkeitsinformationen Soluble in methanol. Insoluble in water.
Formelmasse 209.2
Reinheit (%) ≥98%
Menge 25 g