missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-4-trans-Hydroxy-L-Prolin-Methylester, 98 %, Thermo Scientific Chemicals

Catalog Number 15438626
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
64187-48-0
C14H17NO5
279.29
MFCD00055851,MFCD09954933,MFCD09954934
VVKAGQHUUDRPOI-UHFFFAOYNA-N
z-hyp-ome, 2s,4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-l-4-hydroxyproline methyl ester, 1-benzyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-trans-4-hydroxy-l-proline methyl ester, trans-n-cbz-4-hydroxy-l-proline methyl ester, l-zhprome, z-l-hyp-ome, z-hydroxy-l-proline methyl ester, 1,2-pyrrolidinedicarboxylicacid, 4-hydroxy-, 2-methyl 1-phenylmethyl ester, 2s,4r
688411
1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidin-1,2-dicarboxylat
COC(=O)C1CC(O)CN1C(=O)OCC1=CC=CC=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
64187-48-0
C14H17NO5
279.29
MFCD00055851,MFCD09954933,MFCD09954934
VVKAGQHUUDRPOI-UHFFFAOYNA-N
z-hyp-ome, 2s,4r-1-benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-l-4-hydroxyproline methyl ester, 1-benzyl 2-methyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, n-cbz-trans-4-hydroxy-l-proline methyl ester, trans-n-cbz-4-hydroxy-l-proline methyl ester, l-zhprome, z-l-hyp-ome, z-hydroxy-l-proline methyl ester, 1,2-pyrrolidinedicarboxylicacid, 4-hydroxy-, 2-methyl 1-phenylmethyl ester, 2s,4r
688411
1-O-benzyl 2-O-methyl (2S,4R)-4-hydroxypyrrolidin-1,2-dicarboxylat
COC(=O)C1CC(O)CN1C(=O)OCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester
Farbe Gelb
Formelmasse 279.29
Reinheit (%) 98%
Menge 250 mg
Physikalische Form Flüssig

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'