Learn More
Thermo Scientific™ N-Acetyl-Ile-Glu-Thr-Asp-7-amino-4-(trifluoromethyl)cumarin
Marke: Thermo Scientific™ J64258.#0
Additional Details : CAS : 211990-57-7 Gewicht : 0.00005kg
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
5 mg | |
-30°C to -10°C | |
C31H38F3N5O12 | |
Ac-IETD-AFC; | |
N-Acetyl-Ile-Glu-Thr-Asp-7-amino-4-(trifluoromethyl)coumarin |
White to off-white | |
Ein fluorogenes Substrat für Caspasen-6 (Mch2), -8 (FLICE), -10 (Mch4) und andere verwandte Cystein-Proteasen. Auch ein Substrat für Granzym B. | |
729.65 | |
Soluble in DMSO | |
Lyophilisiertes Pulver |